Learn More
Neuromics, Inc Charcoal Stripped Fetal Bovine Serum, 500 ml removes nonpolar molecules, including viruses, certain growth factors, hormones, and cytokines, without removing nutrients.
Catalog No. NC2071930
Encompass
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability and to ensure product safety and performance.Our Exosome Depleted FBS features: Removal of nonpolar molecules, Triple 0.1 μm Filtered, Free of Virus and Mycoplasma.Charcoal Stripped FBS removes nonpolar molecules (viruses, certain growth factors, hormones, cytokines) without removing nutrients (salts, glucose, amino acids). Excellent for research involving hormones, cancer, cell signaling, drug response, etc.
Catalog No. NC2071930 Supplier Neuromics, Inc Supplier No. FBS005C
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability and to ensure product safety and performance.Our Exosome Depleted FBS features: Removal of nonpolar molecules, Triple 0.1 μm Filtered, Free of Virus and Mycoplasma.Charcoal Stripped FBS removes nonpolar molecules (viruses, certain growth factors, hormones, cytokines) without removing nutrients (salts, glucose, amino acids). Excellent for research involving hormones, cancer, cell signaling, drug response, etc.