Learn More
Med Vet International Yellow Charged Slides, 10gr/case
Catalog No. 503045333
Encompass_Preferred
Special adhesive formula coated on slides also known as Positively Charged allows the specimen to adhere to slide easily, saving time and money in lab procedures. Packed 72pcs/box, 20 boxes (10 gross) per case. Order per gross in multiples of 10.
Catalog No. 50-304-5333 Supplier Med Vet International Supplier No. 9308Y
Special adhesive formula coated on slides also known as Positively Charged allows the specimen to adhere to slide easily, saving time and money in lab procedures. Packed 72pcs/box, 20 boxes (10 gross) per case. Order per gross in multiples of 10.