Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Charger for EVAHL (Hazardous Locations) PAPR Systems
SDP

Catalog No. 17989544 Shop All Bullard Company Products
Change view
Click to view available options
No. of Ports:
One
Six
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Ports
17-989-544 Six
17-989-542 One
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-989-544 Supplier Bullard™ Supplier No. EVAHLGC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator (PAPR).

  • Choice of single or six port configurations to suit most needs

Specifications

Color Black
Product Type Multi-Unit Charging Station
For Use With (Equipment) EVAHL PAPR Systems
Model EVAHLGC
No. of Ports Six

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.