Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChargeSwitch™ gDNA Rendered Meat Purification Kit

Catalog No. CS400100
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CS400100 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CS400100 Supplier Invitrogen™ Supplier No. CS400100
Add to Cart
Add to Cart

The ChargeSwitch™ gDNA Rendered Meat Purification Kit allows for rapid and efficient purification of genomic DNA (gDNA) from animal feed, animal meal, and heparin products for downstream molecular analysis.

The ChargeSwitch™ gDNA Rendered Meat Purification Kit allows for rapid and efficient purification of genomic DNA (gDNA) from animal feed, animal meal, and heparin products for downstream molecular analysis. The kit is listed in the U.S. Food and Drug Administration Center for Veterinary Medicine's protocols as a sample preparation method suitable for monitoring crude heparin quality and testing animal feed for banned materials such as specific animal proteins.

Note: The number of reactions possible with this kit is dependent upon the starting sample type: animal feed and meal samples = 50 preparations, heparin products = 25 preparations.

The ChargeSwitch™ gDNA Rendered Meat Purification Kit protocol starts with prepared lysates and takes approximately 15 minutes to complete without centrifugation, vacuum, or organic solvents (for batches of up to 5 samples per operator—larger batch sizes will require longer processing times). The kit employs a magnetic bead with a switchable surface. Low pH conditions cause binding of DNA to the bead while high pH conditions cause the release of the DNA.

Tested by the U.S. Food and Drug Administration

The ChargeSwitch™ gDNA Rendered Meat Purification Kit has been thoroughly tested by the U.S. Food and Drug Administration for processing up to 500 mg of animal feed, 250 mg of animal meal, and 250 mg of heparin product per reaction.

References

Development of a Multiplex Real-Time PCR Assay for the Detection of Ruminant DNA in Raw Materials used for Monitoring Crude Heparin for Quality: www.fda.gov/AnimalVeterinary/ScienceResearch/ToolsResources/ucm350289.html

Development, evaluation, and peer verification of a rapid real-time PCR method for the detection of animal material. Journal of Food Protection, Vol. 72, No. 11, 2009, Pages 2368–2374. U.S. Food and Drug Administration, Center for Veterinary Medicine, Office of Research, Laurel, Maryland 20708, USA.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Magnetic Beads, 1.1 mL
• SDS Solution, 10 mL
• 10% Detergent, 10 mL
• Lysis Buffer, 100 mL
• Precipitation Buffer, 38.5 mL
• Wash Buffer, 200 mL
• Elution Buffer, 15 mL
Isolation Technology Magnetic Bead
Sample Type Biopharmaceutical Production Samples, Food and Environmental
For Use With (Application) Real-Time Quantitative PCR (qPCR), PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 50 Reactions
Product Line ChargeSwitch
Quantity 50 reactions
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.