Learn More
Active Motif Chariot™
SDP
Catalog No. NC9766803
Encompass
Chariot is a revolutionary delivery reagent that quickly and efficiently transports biologically active proteins, peptides and antibodies directly into cells. The typical delivery efficiency is 60-95%. Less than two hours after delivery, live cells can be assayed to determine the effects of the introduced material, without the need for fixing. This makes Chariot an ideal tool for functional studies, including delivering inhibitory proteins, labeling organelles, screening peptide libraries and studying protein half-lives & transient complementation.
Catalog No. NC9766803 Supplier Active Motif Supplier No. 30025
Chariot is a revolutionary delivery reagent that quickly and efficiently transports biologically active proteins, peptides and antibodies directly into cells. The typical delivery efficiency is 60-95%. Less than two hours after delivery, live cells can be assayed to determine the effects of the introduced material, without the need for fixing. This makes Chariot an ideal tool for functional studies, including delivering inhibitory proteins, labeling organelles, screening peptide libraries and studying protein half-lives & transient complementation.