Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Charles River Axcess™ System Qualification Panel
SDP

Catalog No. 23045547 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
30 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-547 30 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-045-547 Supplier Microbiologics™ Supplier No. 5227P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics offers a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.

Charles River Axcess™ System Qualification Panel contains two KWIK-STIKs each Bacillus subtilis subsp. subtilis derived from ATCC™ 6051™, Brevundimonas vesicularis derived from ATCC™ 11426™,Candida albicans derived from ATCC™ 14053™,Chryseobacterium indologenes derived from ATCC™ 29897™, Curtobacterium pusillum derived from ATCC™ 19096™,Deinococcus radiophilus derived from ATCC™ 27603™,Enterococcus faecalis derived from ATCC™ 19433™, Methylobacterium organophilum derived from ATCC™ 27886™,Micrococcus luteus ;derived from ATCC™ 4698™,Proteus vulgaris derived from ATCC™ 29905™,Ralstonia pickettii derived from ATCC™ 27511™,Pseudomonas aeruginosa derived from ATCC™ 10145™,Staphylococcus aureus subsp. aureus derived from ATCC™ 12600™,Staphylococcus epidermidis derived from ATCC™ 14990™, and Stenotrophomonas maltophilia derived from ATCC™ 13637™

Specifications

Content And Storage 2°C to 8°C
Format QC Sets and Panels: KWIK-STIK™
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Microbial Identification
No. per Pack 2
Packaging Type Plastic Canister
Quantity 30 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.