Learn More
LESMAN INSTRUMENT COMPANY 10-5/16" 7D Circular Chart, 100/box
Catalog No. NC9723496
Encompass_Preferred
Honeywell Circular Chart | Chart Size (Diam): 10-5/16-Inch | 7 Day Rotation | Range: Volts DC: 0 to 5V | Chart Calibration: 0 to 50 | Compatible Recorders: AR100, DR4200, DR4300 | Box of 100.
Catalog No. NC9723496 Supplier LESMAN INSTRUMENT COMPANY Supplier No. 24001661051
Honeywell Circular Chart | Chart Size (Diam): 10-5/16-Inch | 7 Day Rotation | Range: Volts DC: 0 to 5V | Chart Calibration: 0 to 50 | Compatible Recorders: AR100, DR4200, DR4300 | Box of 100.