Learn More
Bellco Glass, Inc. Temp Chart Recorder,115V 7Day 23Hr
SDP
Catalog No. 501946048
Encompass_Preferred
Optional temperature chart recorder for Roll-In Incubators. The chart recorder records up to 24 hours a day and 7 days a week. The unit is housed in a grey plastic case with a 6" viewing window. Users can select temperature ranges (preset to 0-100C) & audible alarm is included. Recorder includes wall mount power adapter & type k thermocouple with 4' bead wire probe plus 50 chart paper circles. Recorder may be wall mounted or panel mounted.NOTE: can be factory installed with r/in inc only for additional pricing. MADE IN THE USA
Catalog No. 50-194-6048 Supplier Bellco Glass, Inc. Supplier No. 772835115
Optional temperature chart recorder for Roll-In Incubators. The chart recorder records up to 24 hours a day and 7 days a week. The unit is housed in a grey plastic case with a 6" viewing window. Users can select temperature ranges (preset to 0-100C) & audible alarm is included. Recorder includes wall mount power adapter & type k thermocouple with 4' bead wire probe plus 50 chart paper circles. Recorder may be wall mounted or panel mounted.NOTE: can be factory installed with r/in inc only for additional pricing. MADE IN THE USA