Learn More
Graphic Controls LLC CHART PAPER HELMER 52 SHEETS
Catalog No. NC0928490
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Chart Size: 04.500" Time Rotation: 7D Number of Ranges: 1 Ranges: (-50) - (0), Uniform along the Arc -40 TO 10 BY 10
Catalog No. NC0928490 Supplier Graphic Controls LLC Supplier No. 32015341
This item has a minimum qty of 2 per supplier requirements.
Chart Size: 04.500" Time Rotation: 7D Number of Ranges: 1 Ranges: (-50) - (0), Uniform along the Arc -40 TO 10 BY 10