Learn More
Cobex Recorders Inc Chart Recorder Paper, Inkless, 6-inch, 7 Day, 50/Pack
SDP
Supplier:  Cobex Recorders Inc 220064
Encompass_Preferred
6 in. circular recording chart paper, inkless style, -5/+50 scale, 1 deg. per division, 7 day rotation, 220064
Catalog No. NC0692805