Learn More
Thermo Scientific™ Chart Recorders for Thermo Scientific™ Ultra-Low Temperature Freezers
Catalog No. CRP4567 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Forma 88000, HERAfreeze HFU-T, Revco UxF and TSU Series (not available on 14.9 cu. ft. freezers)
Catalog No. For Use With (Equipment)
CRP4567 Forma 88000, HERAfreeze HFU-T, Revco UxF and TSU Series (not available on 14.9 cu. ft. freezers)
1 options
Catalog No. CRP4567 Supplier Thermo Scientific™ Supplier No. CRP4567
Add to Cart
Add to Cart
For use in Ultra-Low Temperature Freezers
  • Seven-day recorder
  • Functions to -150°C
  • Field† or Factory installed
Field-installed units must be installed by a qualified professional
TRUSTED_SUSTAINABILITY
Specifications
Type Temperature Recorder
For Use With (Equipment) Forma 88000, HERAfreeze HFU-T, Revco UxF and TSU Series (not available on 14.9 cu. ft. freezers)