Learn More
Alkali Scientific Chart™ Trifecta Ambient Vaporizers – VAP AMB AL, 10K
SDP
Supplier:  Alkali Scientific FG02667
Encompass_Preferred
The Chart™ Trifecta Ambient Vaporizers are engineered for efficient, reliable vaporization of CO₂ at ambient temperatures. Designed for industrial and commercial applications, these vaporizers deliver consistent gas flow and pressure without the need for external heating, ensuring optimal performance and energy efficiency. Built with durable, corrosion resistant materials, they offer long lasting operation, simplified maintenance, and enhanced safety, making them ideal for demanding environments where dependable CO₂ vaporization is critical. Key Features: Efficient Ambient Vaporization: Delivers consistent CO₂ flow without external heating. Reliable Performance: Maintains stable pressure for industrial and commercial applications. Durable Construction: Corrosion resistant materials ensure long term operation. Energy Efficient Design: Optimizes performance while minimizing energy use. Enhanced Safety: Built in features protect against overpressure and leaks. Low
Catalog No. 50-006-62984