Learn More
Enterprise Technology Solutions Chartpak Drafting Film, Permanent, Matte, 8-1/2"x11",100/BX
SDP
Catalog No. 5000826497
Encompass_Preferred
This 1.5 mil drafting applique film with matte finish helps save time when any application needs to be drafted or drawn multiple times. The matte surface gives top-quality images using copiers, laser printers, computer printers, typewriters, pencil or ink. Ideal for diazo reproduction. Polyester drafting film will not yellow with age. Use with copiers and printers.
Catalog No. 50-008-26497 Supplier Enterprise Technology Solutions Supplier No. CHADAF8
This 1.5 mil drafting applique film with matte finish helps save time when any application needs to be drafted or drawn multiple times. The matte surface gives top-quality images using copiers, laser printers, computer printers, typewriters, pencil or ink. Ideal for diazo reproduction. Polyester drafting film will not yellow with age. Use with copiers and printers.