Learn More
AdipoGen Chartreusin
SDP
Supplier:  AdipoGen BVT0005M005
Encompass_Preferred
Chemical. CAS 6377-18-0. Formula C32H32O14. MW 640.6. Isolated from Streptomyces chartreusis. Antibiotic Antitumor compound. Topoisomerase II inhibitor. Induces single-strand scission in DNA in the presence of reducing agents. Apoptosis inducer. Inhibitor of glioblastoma multiforme GBM.
Catalog No. 50-256-2156