Learn More
Cayman Chemical A23187, Purity: ≥98%, CAS: 52665-69-7, 10 mg
SDP
Catalog No. 502951396
Encompass_Preferred
A calcium ionophore and a microbial metabolite; induces calcium overload and mitochondrial apoptosis in HEK293 cells at 5 µM; induces the production of ROS and NET formation in primary human neutrophils at 5 µM
Catalog No. 50-295-1396 Supplier Cayman Chemical Supplier No. 1101610 MG
A calcium ionophore and a microbial metabolite; induces calcium overload and mitochondrial apoptosis in HEK293 cells at 5 µM; induces the production of ROS and NET formation in primary human neutrophils at 5 µM