Learn More
Medchemexpress LLC Charybdotoxin TFA | 96.8% | 4295.89 (free acid) | 100 UG
SDP
Catalog No. 5000425039
Encompass_Preferred
Charybdotoxin TFA is a 37-amino acid peptide that functions as a potassium channel blocker. This compound is a valuable tool for research involving potassium channels.
  • Acts as a potassium channel blocker
  • 37-amino acid peptide
  • Useful for studying potassium channels
  • Storage conditions for powder: -80°C for 2 years, -20°C for 1 year
  • Storage conditions for in solvent: -80°C for 6 months, -20°C for 1 month
  • Sealed storage, away from moisture and light required for both forms
Catalog No. 50-004-25039 Supplier Medchemexpress LLC Supplier No. HYP0191A100UG
Charybdotoxin TFA is a 37-amino acid peptide that functions as a potassium channel blocker. This compound is a valuable tool for research involving potassium channels.
  • Acts as a potassium channel blocker
  • 37-amino acid peptide
  • Useful for studying potassium channels
  • Storage conditions for powder: -80°C for 2 years, -20°C for 1 year
  • Storage conditions for in solvent: -80°C for 6 months, -20°C for 1 month
  • Sealed storage, away from moisture and light required for both forms