Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Charybdotoxin (TFA) | 96.8% | 4295.89 (free acid) | 500 UG
SDP

Catalog No. 5000425882
Encompass_Preferred

Charybdotoxin (TFA) is a research-grade peptide supplied as a white to off-white solid. It has a molecular weight of 4295.89 (free acid) and a purity of 96.8% as determined by LCMS. This substance is intended for laboratory use and substance manufacturing, offering stability under recommended storage conditions.

  • Supplied as a white to off-white solid
  • Purity of 96.8% (LCMS)
  • Molecular weight of 4295.89 (free acid)
  • Suitable for use as laboratory chemical and for substance manufacturing
  • Stable under recommended storage conditions
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-25882 Supplier Medchemexpress LLC Supplier No. HYP0191A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Charybdotoxin (TFA) is a research-grade peptide supplied as a white to off-white solid. It has a molecular weight of 4295.89 (free acid) and a purity of 96.8% as determined by LCMS. This substance is intended for laboratory use and substance manufacturing, offering stability under recommended storage conditions.

  • Supplied as a white to off-white solid
  • Purity of 96.8% (LCMS)
  • Molecular weight of 4295.89 (free acid)
  • Suitable for use as laboratory chemical and for substance manufacturing
  • Stable under recommended storage conditions
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.