Learn More
MSC New Pig Absorbent Roll, High-Visibility, 24 in, 1 roll
Catalog No. 5000675441
Encompass
This high-visibility yellow absorbent roll is designed to clean up floor spills and convey safety messages. Featuring a repeating safety sign image, it is suitable for placement in aisles and walkways. Its mediumweight, wide rolls effectively absorb everyday spills, leaks, and overspray around machinery or in large areas.
  • Cleans up spills while sending a clear safety message
  • Bright yellow color ensures high visibility of hazards and cleanups
  • Durable top layer resists scuffs and withstands foot traffic
  • Resists ripping, tearing, or fraying even when saturated
  • Quickly absorbs oils, coolants, solvents, and water
  • Easy-tear perforations allow for custom sizing and waste reduction
  • Can be wrung out and incinerated after use
Catalog No. 50-006-75441 Supplier MSC Supplier No. 83184358
This high-visibility yellow absorbent roll is designed to clean up floor spills and convey safety messages. Featuring a repeating safety sign image, it is suitable for placement in aisles and walkways. Its mediumweight, wide rolls effectively absorb everyday spills, leaks, and overspray around machinery or in large areas.
  • Cleans up spills while sending a clear safety message
  • Bright yellow color ensures high visibility of hazards and cleanups
  • Durable top layer resists scuffs and withstands foot traffic
  • Resists ripping, tearing, or fraying even when saturated
  • Quickly absorbs oils, coolants, solvents, and water
  • Easy-tear perforations allow for custom sizing and waste reduction
  • Can be wrung out and incinerated after use