Learn More
Apexbio Technology LLC CHC 28166-41-8 50mg
SDP
Catalog No. 502240619
Encompass_Preferred
CHC (CAS 28166-41-8) is a derivative of -cyano-4-hydroxycinnamic acid that acts as a non-competitive inhibitor of monocarboxylate transporters (MCTs) thereby modulating monocarboxylate translocation and affecting associated cellular pathways In biomedical research CHC is commonly utilized to study monocarboxylate transporter function and related enzymatic activities Additionally it is widely employed as an auxiliary matrix in matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry where it facilitates peptide ionization supporting analytical and proteomics investigations
This item is not returnable. View return policy
Catalog No. 50-224-0619 Supplier Apexbio Technology LLC Supplier No. B773350
CHC (CAS 28166-41-8) is a derivative of -cyano-4-hydroxycinnamic acid that acts as a non-competitive inhibitor of monocarboxylate transporters (MCTs) thereby modulating monocarboxylate translocation and affecting associated cellular pathways In biomedical research CHC is commonly utilized to study monocarboxylate transporter function and related enzymatic activities Additionally it is widely employed as an auxiliary matrix in matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry where it facilitates peptide ionization supporting analytical and proteomics investigations