Learn More
CPC Scientific H-Arg-Phe-Gly-Arg-Phe-Leu-Arg-Lys Ile-Arg-Arg-Phe-Arg-Pro-Lys-Val-Thr-Ile-Thr-Ile-Gln-Gly-Ser-Ala-Arg Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATMP011A
Encompass
Despite having no homology to peptide LL37, chicken cathelicidin-2 (CATH-2) is a potent antimicrobial peptide. Both chCATH-2 and LL37 exhibit amphipathic alpha-helical conformations.ONE-LETTER SEQUENCE: RFGRFLRKIRRFRPKVTITIQGSARF-NH2MOLECULAR FORMULA: C147H245N51O30MOLECULAR WEIGHT:3206.86STORAGE CONDITIONS: -20 5CSYNONYMS: chicken cathelicidin 2; CMAP27, chicken myeloid antimicrobial peptide 27, Fowlicidin-2; chCATH-2; birds, animals; BBL; Derivatives: F2,5,12WRESEARCH AREA: AntimicrobialREFERENCES: Van Dijk, Albert, et al. Molecular immunology 46.13 (2009): 2465-2473.
Catalog No. 50-210-8826