Learn More
Aviva Systems Biology Corp CHCHD1 Antibody - N-terminal region (ARP53490_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHCHD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHCHD1. Alternate names: C10orf34, C2360, FLJ25854ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-843