Learn More
Aviva Systems Biology Corp CHCHD3 Antibody - N-terminal region (ARP57039_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57039_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHCHD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHCHD3. Alternate names: FLJ20420, MINOS3, PPP1R22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62065