Learn More
Aviva Systems Biology Corp CHCHD4 Antibody - N-terminal region (ARP51895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHCHD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHCHD4. Alternate names: FLJ31709, MIA40, TIMM40ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53677