Learn More
ABclonal Technology CHCHD4 Rabbit pAb
SDP
Catalog No. 5000097918
Encompass_Preferred
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus (Hofmann et al., 2005 [PubMed 16185709]).
Catalog No. 50-000-97918 Supplier ABclonal Technology Supplier No. A1313920UL
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus (Hofmann et al., 2005 [PubMed 16185709]).