Learn More
Aviva Systems Biology Corp CHCHD6 Antibody - N-terminal region (ARP60420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHCHD6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CHCHD6. Alternate names: MGC13016, CHCM1, PPP1R23ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-287-038