Learn More
World Precision Instrument Check Valve, Female Luer Lock Inlet, Male Luer Lock Outlet
SDP
Catalog No. 50822025
Encompass
  • Polyester & Silicone Check Valve
  • 0.170PSI Crack Pressure
  • Package of 10
Catalog No. 50-822-025 Supplier World Precision Instrument Supplier No. 1403910
  • Polyester & Silicone Check Valve
  • 0.170PSI Crack Pressure
  • Package of 10