Learn More
Aviva Systems Biology Corp CHD1 Antibody - middle region (ARP34368_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34368_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHD1. Alternate names: DKFZp686E2337ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-55357