Learn More
Aviva Systems Biology Corp CHD1L Antibody - middle region (ARP36364_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36364_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHD1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHD1L. Alternate names: ALC1, CHDLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56127