Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CHD1L Recombinant Enzymes
SDP

Catalog No. NBP348863
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP348863 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP348863 Supplier Novus Biologicals™ Supplier No. NBP348863

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Recombinant Enzyme

Specifications

Gene ID (Entrez) 9557
Species Human
Purity Immunogen affinity purified
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias ALC1, amplified in liver cancer 1, amplified in liver cancer protein 1, CHDL, chromodomain helicase DNA binding protein 1-like, chromodomain-helicase-DNA-binding protein 1-like, FLJ22530
Gene Symbol CHD1L
Quantity 50 μL
Storage Buffer 20 mM Tris-HCl, pH 7.9, 150 mM NaCl, 15% glycerol, 2 mM MgCl2, 0.2 mM EDTA, 1 mM DTT, 10 mM beta-glycerophospate, 0.4 mM PMSF, 1 mM benzamidine and 0.4 mg/mL recombinant human insulin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.