Learn More
ABclonal Technology CHDH Rabbit mAb
SDP
Catalog No. 5000010655
Encompass_Preferred
The protein encoded by this gene is a choline dehydrogenase that localizes to the mitochondrion Variations in this gene can affect susceptibility to choline deficiency A few transcript variants have been found for this gene but the full-length nature of only one has been characterized to date
Catalog No. 50-000-10655 Supplier ABclonal Technology Supplier No. A2584320UL
The protein encoded by this gene is a choline dehydrogenase that localizes to the mitochondrion Variations in this gene can affect susceptibility to choline deficiency A few transcript variants have been found for this gene but the full-length nature of only one has been characterized to date