Learn More
Medchemexpress LLC Chdi-390576 | 1629729-98-1 | 98.1% | 25 MG
SDP
Catalog No. 5000234327
Encompass_Preferred
This small-molecule class IIa histone deacetylase (HDAC) inhibitor is reported to be cell-permeable and to penetrate the central nervous system. It is supplied in research pack sizes and analytical formats, with molecular formula C19H13F4N3O2, molecular weight 391.32 g/mol, and reported purity 98.10%.
  • Potent inhibition of class IIa HDACs (IC50s: 54 nM, 60 nM, 31 nM, 50 nM).
  • Cell permeable and CNS penetrant.
  • Available in multiple pack sizes and solution formats.
  • High reported purity suitable for research assays.
  • Analytical documentation available (COA, SDS, HNMR, LCMS, RP-HPLC).
Catalog No. 50-002-34327 Supplier Medchemexpress LLC Supplier No. HY11993925MG
This small-molecule class IIa histone deacetylase (HDAC) inhibitor is reported to be cell-permeable and to penetrate the central nervous system. It is supplied in research pack sizes and analytical formats, with molecular formula C19H13F4N3O2, molecular weight 391.32 g/mol, and reported purity 98.10%.
  • Potent inhibition of class IIa HDACs (IC50s: 54 nM, 60 nM, 31 nM, 50 nM).
  • Cell permeable and CNS penetrant.
  • Available in multiple pack sizes and solution formats.
  • High reported purity suitable for research assays.
  • Analytical documentation available (COA, SDS, HNMR, LCMS, RP-HPLC).