Learn More
Phoenix Flow Systems Inc CHEAP SHEATH PBS CONCENTRATE
Catalog No. NC9063042
Encompass
Cheap Sheath Is A Concentrated Ph Balanced Formulation Of Phosphate Buffered Saline Which Has Been Pre-Filtered And Is Ready To Use In Any Flow Cytometer As Your Sheath Solution Upon Dilution With Distilled Water. Each Bottle Of Cheap Sheath Contains Enough Concentrate To Prepare 20 Liters Of Flow Cytometer Sheath Soluton. Formulation Is Simple For Flow Cytometers That Do Not Use 20 Liter Sheath Containers. For Any Flow Cytometer With A 3.5 Liter Sheath Tank, Add 100 Mls Cheap Sheath To 2.9 Liters Of Distilled Water In The Sheath Tank. 1Qt Bottle (2 Lbs).
Catalog No. NC9063042 Supplier Phoenix Flow Systems Inc Supplier No. CS001
Add to Cart
Add to Cart
Cheap Sheath Is A Concentrated Ph Balanced Formulation Of Phosphate Buffered Saline Which Has Been Pre-Filtered And Is Ready To Use In Any Flow Cytometer As Your Sheath Solution Upon Dilution With Distilled Water. Each Bottle Of Cheap Sheath Contains Enough Concentrate To Prepare 20 Liters Of Flow Cytometer Sheath Soluton. Formulation Is Simple For Flow Cytometers That Do Not Use 20 Liter Sheath Containers. For Any Flow Cytometer With A 3.5 Liter Sheath Tank, Add 100 Mls Cheap Sheath To 2.9 Liters Of Distilled Water In The Sheath Tank. 1Qt Bottle (2 Lbs).