Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Flow Systems Inc CHEAP SHEATH START PACK 5/PK

Catalog No. NC1530523
Encompass

  • Flow cytometer sheath fluid concentrate
  • pH balanced phosphate buffered saline (PBS)
  • Pre-filtered concentrate for purity
  • Pack contains 5 concentrate bottles and 1 reusable 20 L container
  • 2 lbs per bottle of concentrate
  • 20 L sheath fluid yield per bottle
  • 100 L total sheath fluid yield per pack
  • Diluted with distilled water
  • Application in flow cytometry
  • Reduced space requirements compared to bulky cubitainers
  • Lighter weight for shipping
  • Environmentally friendly with less landfill waste
  • Simple to prepare for easy use

Catalog No. NC1530523 Supplier Phoenix Flow Systems Inc Supplier No. CSSTARTERPACK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Flow cytometer sheath fluid concentrate
  • pH balanced phosphate buffered saline (PBS)
  • Pre-filtered concentrate for purity
  • Pack contains 5 concentrate bottles and 1 reusable 20 L container
  • 2 lbs per bottle of concentrate
  • 20 L sheath fluid yield per bottle
  • 100 L total sheath fluid yield per pack
  • Diluted with distilled water
  • Application in flow cytometry
  • Reduced space requirements compared to bulky cubitainers
  • Lighter weight for shipping
  • Environmentally friendly with less landfill waste
  • Simple to prepare for easy use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.