Learn More
Medchemexpress LLC Chebulic acid | 23725-05-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN41701MG
Encompass_Preferred
Chebulic acid is a phenolic acid compound isolated from the plant *Terminalia chebula*, recognized for its strong antioxidant properties. It functions by disrupting protein cross-links caused by advanced glycation end products (AGEs) and preventing the formation of new AGEs. This compound has demonstrated effectiveness in addressing elevated metabolic parameters, oxidative stress, and liver damage, indicating its potential therapeutic role in conditions such as asthma, diabetes, and for general liver protection. It is intended for research use only.
  • Potent antioxidant activity
  • Disrupts and inhibits advanced glycation end products (AGEs)
  • Effective in managing elevated metabolic parameters
  • Reduces oxidative stress
  • Supports liver health and protection
  • Potential therapeutic agent for asthma and diabetes research
Catalog No. 50-004-20847