Learn More
Medchemexpress LLC Chebulic acid | 23725-05-5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN417010MM/1ML
Encompass_Preferred
Chebulic acid is a phenolic acid compound isolated from *Terminalia chebula* with strong antioxidant activity. It breaks protein cross-links induced by advanced glycation end products (AGEs) and inhibits the formation of AGEs. Chebulic acid is effective in controlling elevated metabolic parameters, oxidative stress, and liver damage, supporting its beneficial role in asthma, diabetes, and liver protection.
  • Isolated from *Terminalia chebula*.
  • Exhibits strong antioxidant activity.
  • Breaks protein cross-links induced by advanced glycation end products (AGEs).
  • Inhibits advanced glycation end product formation.
  • Helps control elevated metabolic parameters, oxidative stress, and liver damage.
  • Supports beneficial roles in asthma and diabetes.
  • Features a high purity of 99.89%.
Catalog No. 50-004-20845