Learn More
Medchemexpress LLC HY-N2033 5mg Medchemexpress, Chebulinic acid CAS:18942-26-2 Purity:>98%
SDP
Catalog No. 501970360
Encompass_Preferred
Medchemexpress, HY-N2033 5mg Chebulinic acid CAS:18942-26-2 Chebulinic acid is a potent natural inhibitor of M. tuberculosis DNA gyrase, also can inhibit SMAD-3 phosphorylation, inhibit H+ K+-ATPase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0360 Supplier Medchemexpress LLC Supplier No. HYN20335MG
Medchemexpress, HY-N2033 5mg Chebulinic acid CAS:18942-26-2 Chebulinic acid is a potent natural inhibitor of M. tuberculosis DNA gyrase, also can inhibit SMAD-3 phosphorylation, inhibit H+ K+-ATPase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.