Learn More
YSI 3P LifeScience CHECK SOLUTION 100MG/L
Catalog No. NC9267186
Encompass
100 mg/L Ammonium and 200 mg/L Potassium Standard Solution for use with YSI Analyzers.
Catalog No. NC9267186 Supplier YSI 3P LifeScience Supplier No. 7179
100 mg/L Ammonium and 200 mg/L Potassium Standard Solution for use with YSI Analyzers.