Learn More
Arkray Check Strip Set (White/Gray)
SDP
Catalog No. 21403492
Change view
Click to view available options
Quantity:
2 Strips/Vial
Catalog No. Quantity
21-403-492 2 Strips/Vial
1 options
For in vitro diagnostic use only. Use only with compatible ARKRAY analyzers and according to the Instructions for Use. Improper use may affect verification results.
Catalog No. 21-403-492 Supplier Arkray Supplier No. 100457
Add to Cart
Add to Cart
For in vitro diagnostic use only. Use only with compatible ARKRAY analyzers and according to the Instructions for Use. Improper use may affect verification results.
The Check Strip Set (White/Gray) by ARKRAY supports quick and reliable instrument verification, helping ensure accurate test performance. Designed for easy use, it streamlines quality checks and supports consistent, confident daily operation.
  • Designed for instrument verification: The Check Strip Set (White/Gray) - is used to verify analyzer performance, helping confirm that systems are operating within expected parameters.
  • Supports quality assurance: Assists laboratories in maintaining consistent testing accuracy by providing a simple, reliable check as part of routine quality processes.
  • Quick and easy to use: Simple application allows staff to perform checks efficiently with minimal training, reducing workflow disruption.
  • Distinct white and gray strips: Color-coded design helps ensure proper identification and use, minimizing handling errors during verification procedures.
  • Promotes confidence in results: Regular use helps reinforce trust in instrument readiness before patient or sample testing begins.
  • Optimized for ARKRAY systems: Specifically designed to work seamlessly with compatible ARKRAY analyzers for reliable and consistent performance checks.
  • Streamlines daily operations: Integrates easily into routine lab workflows without adding unnecessary complexity or time.
  • Consistent performance standards: Manufactured to meet ARKRAY quality specifications, supporting dependable verification from strip to strip.
  • Ideal for routine use: Suitable for daily, scheduled, or as-needed checks to support ongoing instrument reliability.
  • Supports operational efficiency: Helps reduce downtime by enabling early identification of potential performance issues.
Specifications
CE Marker CE-IVD
Clia Complexity High, Moderate
Type Test Strips
Disposable Single-use
Quantity 2 Strips/Vial
Form Ready-to-Use (RTU)
The Check Strip Set (White/Gray) is intended for routine instrument verification and quality support as part of regular laboratory workflows.