Learn More
Electron Microscopy Sciences Check Temp 4 Complete
SDP
Catalog No. 501899743
Encompass_Preferred
Checktemp 4 thermometer is fast responding, fold-away probe is made of high quality, stainless steel. It enables angled measurements and can penetrate semi-frozen and semi-solid samples. The LCD on the side of the handle is easy to see and read.
Unfold the stainless steel probe, the Checktemp 4 automatically turns ON and immediately performs a calibration test and also lets you know if your battery level is low or if your meter requires recalibration. These unique features provide the security of knowing you have accurate measurements. It also features auto shut-off.
Catalog No. 50-189-9743 Supplier Electron Microscopy Sciences Supplier No. 7208602
Checktemp 4 thermometer is fast responding, fold-away probe is made of high quality, stainless steel. It enables angled measurements and can penetrate semi-frozen and semi-solid samples. The LCD on the side of the handle is easy to see and read.
Unfold the stainless steel probe, the Checktemp 4 automatically turns ON and immediately performs a calibration test and also lets you know if your battery level is low or if your meter requires recalibration. These unique features provide the security of knowing you have accurate measurements. It also features auto shut-off.