Learn More
COLDER PRODUCTS COMPANY Check Valve 1/2 FNPT X 1/2 MNPT EPR O-Ring 302 Stainless Spring (DrumQuik PRO)
Catalog No. 503131389
Encompass_Preferred
Check Valve 1/2 FNPT X 1/2 MNPT EPR O-Ring 302 Stainless Spring (DrumQuik PRO)
Catalog No. 50-313-1389 Supplier COLDER PRODUCTS COMPANY Supplier No. 2478900
Check Valve 1/2 FNPT X 1/2 MNPT EPR O-Ring 302 Stainless Spring (DrumQuik PRO)