Learn More
Grainger Pneumadyne Inline Poppet Check Valve, Brass, 1/8 in FNPT, 0.5 psi Cracking Pressure
Catalog No. 502750779
Encompass_Preferred
This inline poppet check valve is designed for use in pneumatic circuits that require free flow in one direction and no flow in the opposite direction. It features a brass body with electroless nickel plating for enhanced corrosion resistance, suitable for general purpose applications.
  • Inline poppet mechanism with a single flow design
  • 1/8 in. FNPT x 1/8 in. FNPT connection type
  • 0.5 psi cracking pressure for low-pressure operation
  • Operating pressure range of 0 to 125 psi
  • Media temperature range of -20 °F to +160 °F
  • Buna-N seal material
  • Flow coefficient Cv of 0.41
Catalog No. 50-275-0779 Supplier Grainger Supplier No. 14F301
This inline poppet check valve is designed for use in pneumatic circuits that require free flow in one direction and no flow in the opposite direction. It features a brass body with electroless nickel plating for enhanced corrosion resistance, suitable for general purpose applications.
  • Inline poppet mechanism with a single flow design
  • 1/8 in. FNPT x 1/8 in. FNPT connection type
  • 0.5 psi cracking pressure for low-pressure operation
  • Operating pressure range of 0 to 125 psi
  • Media temperature range of -20 °F to +160 °F
  • Buna-N seal material
  • Flow coefficient Cv of 0.41