Learn More
New Jersey Fluid Systems LLC CHECK VALVE-1/4" W/ 1LB SPRING
Catalog No. NC0824421
Encompass
316 Stainless Steel Swagelok C series Check Valve. 1/4 in. OD Tube Fitting ends. 3,000psi max. 1PSI Spring. Fluorocarbon FKM o-ring
Catalog No. NC0824421 Supplier New Jersey Fluid Systems LLC Supplier No. SS4C1
316 Stainless Steel Swagelok C series Check Valve. 1/4 in. OD Tube Fitting ends. 3,000psi max. 1PSI Spring. Fluorocarbon FKM o-ring