Learn More
Waters Corp Check Valve, 3/8 in., 10000 PSI, High Flow
Catalog No. 502391435 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This check valve is designed for high-flow applications with a P350 pump. It features a 3/8 in. diameter and is rated for 10000 PSI.
  • High-flow check valve
  • Designed for P350 pump
  • 3/8 in. diameter
  • 10000 PSI pressure rating
  • Sold and shipped only within the US to end users
Catalog No. 50-239-1435 Supplier Waters Corp Supplier No. 700006835
This check valve is designed for high-flow applications with a P350 pump. It features a 3/8 in. diameter and is rated for 10000 PSI.
  • High-flow check valve
  • Designed for P350 pump
  • 3/8 in. diameter
  • 10000 PSI pressure rating
  • Sold and shipped only within the US to end users