Learn More
Waters Corp Check Valve, Accumulator, Double Ball and Seat, for UPLC/HPLC Systems
Catalog No. 502390625 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Double ball and seat check valve
  • Designed for use in HPLC and UPLC systems
  • Compatible with ACQUITY UPLC ISM, BSM, QSM, and H-Class QSM components
  • Compatible with Alliance iS HPLC, ACQUITY Arc HPLC, ACQUITY Arc Premier, ACQUITY UPLC H-Class, I-Class, M-Class, and ACQUITY 2D systems
  • Supplied as one unit per package
Catalog No. 50-239-0625 Supplier Waters Corp Supplier No. 700005415
  • Double ball and seat check valve
  • Designed for use in HPLC and UPLC systems
  • Compatible with ACQUITY UPLC ISM, BSM, QSM, and H-Class QSM components
  • Compatible with Alliance iS HPLC, ACQUITY Arc HPLC, ACQUITY Arc Premier, ACQUITY UPLC H-Class, I-Class, M-Class, and ACQUITY 2D systems
  • Supplied as one unit per package