Learn More
Grainger Colorflow C Series Check Valve, Brass, 1/4 In. NPT, 1 ea
Catalog No. 502794277
Encompass_Preferred
These C Series check valves are designed for reliable unidirectional flow control, allowing free flow in one direction and ensuring a secure, leak-tight shut-off in the reverse direction. Engineered for durability, these brass valves are suitable for air, water, and hydraulic oil service. The internal design features a poppet guided by a triangular retainer, which maintains spring stability even under high velocity or shock conditions.
  • Maximum operating pressure of 2,000 psi
  • Temperature range of -40°F to 250°F (-40°C to 121°C)
  • In-line mounting configuration with 1/4 in. NPT connection ports
  • Standard 5 psi cracking pressure
  • Constructed with a brass body, nitrile seals, and stainless steel internal components
  • Maximum flow rate of 19 L/min (5 gpm)
  • Includes soft seal poppet design
Catalog No. 50-279-4277 Supplier Grainger Supplier No. 1A858
These C Series check valves are designed for reliable unidirectional flow control, allowing free flow in one direction and ensuring a secure, leak-tight shut-off in the reverse direction. Engineered for durability, these brass valves are suitable for air, water, and hydraulic oil service. The internal design features a poppet guided by a triangular retainer, which maintains spring stability even under high velocity or shock conditions.
  • Maximum operating pressure of 2,000 psi
  • Temperature range of -40°F to 250°F (-40°C to 121°C)
  • In-line mounting configuration with 1/4 in. NPT connection ports
  • Standard 5 psi cracking pressure
  • Constructed with a brass body, nitrile seals, and stainless steel internal components
  • Maximum flow rate of 19 L/min (5 gpm)
  • Includes soft seal poppet design