Learn More
Grainger Parker Hannifin Colorflow Hydraulic Check Valve, Brass, 1/8 in., 2,000 psi, 3 gpm, 1 ea.
Catalog No. 502794489
Encompass_Preferred
The C Series check valve is designed for hydraulic systems requiring unidirectional flow and reliable reverse shut-off. Featuring a robust brass body, this mechanical valve automatically opens when system pressure reaches the 5 psi cracking threshold and seals tightly when pressure drops below that level.
  • Brass construction with nitrile seals for durability and performance
  • Rated for operating pressures up to 2,000 psi
  • Compatible with hydraulic oil, hydraulic fluids, water, and air
  • Maximum flow rate capacity of 11 L/min (3 gpm)
  • Operating temperature range of -40°C to +121°C (-40°F to 250°F)
  • Compact in-line mounting configuration with 1/8 in. NPT connection
Catalog No. 50-279-4489 Supplier Grainger Supplier No. 1A857
The C Series check valve is designed for hydraulic systems requiring unidirectional flow and reliable reverse shut-off. Featuring a robust brass body, this mechanical valve automatically opens when system pressure reaches the 5 psi cracking threshold and seals tightly when pressure drops below that level.
  • Brass construction with nitrile seals for durability and performance
  • Rated for operating pressures up to 2,000 psi
  • Compatible with hydraulic oil, hydraulic fluids, water, and air
  • Maximum flow rate capacity of 11 L/min (3 gpm)
  • Operating temperature range of -40°C to +121°C (-40°F to 250°F)
  • Compact in-line mounting configuration with 1/8 in. NPT connection