Learn More
Grainger Colorflow C Series Check Valve, Brass, 3/8 in NPT, 5 psi Cracking Pressure, 1 ea
Catalog No. 502794580
Encompass_Preferred
The C Series Colorflow check valve is a high-performance in-line hydraulic check valve designed for reliable directional flow control in hydraulic and pneumatic systems. Constructed from durable brass with a nitrile seal, this valve provides free flow in one direction and ensures tight sealing with minimal leakage. It is suitable for diverse industrial and mobile applications, including accumulator circuits and power units.
  • Brass body construction for corrosion resistance
  • Soft seat poppet design minimizes internal leakage
  • 5 psi cracking pressure
  • 3/8 in. NPT female x 3/8 in. NPT female connection
  • Maximum operating pressure of 2,000 psi
  • Maximum flow rate of 8 gpm (30 L/min)
  • Compatible with hydraulic oil, hydraulic fluids, water, and air
  • Wide operating temperature range from -40°F to 250°F
  • Compact in-line configuration for versatile installation
Catalog No. 50-279-4580 Supplier Grainger Supplier No. 1A859
The C Series Colorflow check valve is a high-performance in-line hydraulic check valve designed for reliable directional flow control in hydraulic and pneumatic systems. Constructed from durable brass with a nitrile seal, this valve provides free flow in one direction and ensures tight sealing with minimal leakage. It is suitable for diverse industrial and mobile applications, including accumulator circuits and power units.
  • Brass body construction for corrosion resistance
  • Soft seat poppet design minimizes internal leakage
  • 5 psi cracking pressure
  • 3/8 in. NPT female x 3/8 in. NPT female connection
  • Maximum operating pressure of 2,000 psi
  • Maximum flow rate of 8 gpm (30 L/min)
  • Compatible with hydraulic oil, hydraulic fluids, water, and air
  • Wide operating temperature range from -40°F to 250°F
  • Compact in-line configuration for versatile installation