Learn More
Waters Corp Waters Check Valve Capsule, HPLC, 1/Package
Catalog No. 50792324 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Check Valve Capsule is designed to simplify check valve replacement in HPLC systems. This capsule contains a ball and seat mechanism, making it suitable for use as both an inlet and outlet valve within the Reagent Manager system.
  • Designed for easy check valve replacement.
  • Contains a ball and seat mechanism.
  • Functions as an inlet or outlet valve in the Reagent Manager system.
  • Includes one unit per package.
  • Compatible with HPLC systems.
  • Recommended for replacement as required.
Catalog No. 50-792-324 Supplier Waters Corp Supplier No. 700000175
The Check Valve Capsule is designed to simplify check valve replacement in HPLC systems. This capsule contains a ball and seat mechanism, making it suitable for use as both an inlet and outlet valve within the Reagent Manager system.
  • Designed for easy check valve replacement.
  • Contains a ball and seat mechanism.
  • Functions as an inlet or outlet valve in the Reagent Manager system.
  • Includes one unit per package.
  • Compatible with HPLC systems.
  • Recommended for replacement as required.