Learn More
Waters Corp Waters Check Valve Cartridge, Ceramic, 2/pk, for HPLC System
Catalog No. 50792801 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These replacement inlet check valve cartridges feature a ball and seat constructed from ceramic. Designed for HPLC systems, this optional check valve can help alleviate sticking issues and features a low internal volume.
  • Replacement inlet check valve cartridges.
  • Ball and seat constructed from ceramic.
  • May help alleviate sticking issues sometimes occurring with other check valves.
  • Low internal volume design.
  • Supplied as 2 units per package.
  • Designed for HPLC systems.
Catalog No. 50-792-801 Supplier Waters Corp Supplier No. 700002761
These replacement inlet check valve cartridges feature a ball and seat constructed from ceramic. Designed for HPLC systems, this optional check valve can help alleviate sticking issues and features a low internal volume.
  • Replacement inlet check valve cartridges.
  • Ball and seat constructed from ceramic.
  • May help alleviate sticking issues sometimes occurring with other check valves.
  • Low internal volume design.
  • Supplied as 2 units per package.
  • Designed for HPLC systems.