Learn More
Waters Corp Waters Check Valve, Double Ball and Seat, UPLC/HPLC Systems
Catalog No. 50793073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement Double Ball & Seat Check Valve designed for Waters ACQUITY UPLC and HPLC systems. It is supplied as a single check valve and is primarily intended for use in an ACQUITY Quaternary Solvent Manager, with compatibility extending to ACQUITY Binary Solvent Manager and ACQUITY UPLC H-Class System.
  • Replacement double ball & seat check valve.
  • Designed for use in a Quaternary Solvent Manager.
  • Compatible with ISM, BSM, and QSM components.
  • Suitable for HPLC and UPLC system types.
  • Has an annual replacement schedule.
Catalog No. 50-793-073 Supplier Waters Corp Supplier No. 700005164
This product is a replacement Double Ball & Seat Check Valve designed for Waters ACQUITY UPLC and HPLC systems. It is supplied as a single check valve and is primarily intended for use in an ACQUITY Quaternary Solvent Manager, with compatibility extending to ACQUITY Binary Solvent Manager and ACQUITY UPLC H-Class System.
  • Replacement double ball & seat check valve.
  • Designed for use in a Quaternary Solvent Manager.
  • Compatible with ISM, BSM, and QSM components.
  • Suitable for HPLC and UPLC system types.
  • Has an annual replacement schedule.